Learn More
Electron Microscopy Sciences MAC Custom Block of 5 Microanalysis Standards
SDP
Supplier:  Electron Microscopy Sciences 8007005
Encompass_Preferred
Pure metal and compounds standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. The standards are polished to a 0.25 micron diamond finish and carbon coated and are supplied with fully authenticated certificates of analyses and a location map for standard identification.
A Faraday Cup, for accurate specimen current measurements is available as an optional extra on all mounts. The standards are mounted in brass holders.
Catalog No. 50-252-17