Learn More
Biotang Inc MAC EWEN SOLUTION, 500 ML,PY-951, 125 mM NaCl, 5 mM KCl, 2 mM CaCl2, 1 mM MgCl2, 8 mM HEPES, 11 mM glucose, pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 0.22 micron filter.
Catalog No. 50152397 Shop All BIOTANG Inc Products
Encompass_Preferred
MAC EWEN SOLUTION, 500 ML,PY-951, 125 mM NaCl, 5 mM KCl, 2 mM CaCl2, 1 mM MgCl2, 8 mM HEPES, 11 mM glucose, pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 0.22 micron filter.
Catalog No. 50-152-397 Supplier Biotang Inc Supplier No. BTBB943
MAC EWEN SOLUTION, 500 ML,PY-951, 125 mM NaCl, 5 mM KCl, 2 mM CaCl2, 1 mM MgCl2, 8 mM HEPES, 11 mM glucose, pH 7.4 +/- 0.2. Prepared in 18.2 megohms water and filtered through 0.22 micron filter.