Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mac glucuronide linker-1 | 2222981-71-5 | 96.4% | 897.90 g/mol | C42H47N3O17S | 1 G
SDP

Catalog No. 5000202917
Encompass_Preferred

MAC glucuronide linker-1 is a glycosidase-cleavable antibody-drug conjugate (ADC) linker supplied as a purified small molecule for research use. It incorporates a glucuronide cleavable moiety to enable enzymatic release of attached payloads, and is provided at high purity for synthesis and conjugation workflows.

  • Glycosidase-cleavable glucuronide linker enabling enzymatic payload release.
  • Molecular weight approximately 897.90 g/mol.
  • Molecular formula C42H47N3O17S.
  • High purity (96.4%) suitable for research applications.
  • Available in multiple sizes including a 1 G package.
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-02917 Supplier Medchemexpress LLC Supplier No. HY442211G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAC glucuronide linker-1 is a glycosidase-cleavable antibody-drug conjugate (ADC) linker supplied as a purified small molecule for research use. It incorporates a glucuronide cleavable moiety to enable enzymatic release of attached payloads, and is provided at high purity for synthesis and conjugation workflows.

  • Glycosidase-cleavable glucuronide linker enabling enzymatic payload release.
  • Molecular weight approximately 897.90 g/mol.
  • Molecular formula C42H47N3O17S.
  • High purity (96.4%) suitable for research applications.
  • Available in multiple sizes including a 1 G package.
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.