Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mac glucuronide linker-1 | 2222981-71-5 | 96.4% | 897.90 g/mol | C42H47N3O17S | 250 MG
SDP

Catalog No. 5000203800
Encompass_Preferred

MAC glucuronide linker-1 is a cleavable ADC linker designed for antibody-agent conjugations and provided for research use. The product is supplied with analytical documentation and recommended storage conditions appropriate for synthesis and conjugation workflows.

  • Cleavable ADC linker for antibody-agent conjugation.
  • CAS number: 2222981-71-5.
  • Molecular weight: 897.90 g/mol.
  • Chemical formula: C42H47N3O17S.
  • Purity (LCMS): 96.36%.
  • Documentation: certificate of analysis and safety data sheet available.
  • Recommended storage: 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple packaging sizes, including 250 mg.

Catalog No. 50-002-03800 Supplier Medchemexpress LLC Supplier No. HY44221250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAC glucuronide linker-1 is a cleavable ADC linker designed for antibody-agent conjugations and provided for research use. The product is supplied with analytical documentation and recommended storage conditions appropriate for synthesis and conjugation workflows.

  • Cleavable ADC linker for antibody-agent conjugation.
  • CAS number: 2222981-71-5.
  • Molecular weight: 897.90 g/mol.
  • Chemical formula: C42H47N3O17S.
  • Purity (LCMS): 96.36%.
  • Documentation: certificate of analysis and safety data sheet available.
  • Recommended storage: 4°C under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple packaging sizes, including 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.