Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mac glucuronide linker-1 | 2222981-71-5 | 96.4% | 897.90 g/mol | C42H47N3O17S | 500 MG
SDP

Catalog No. 5000202616
Encompass_Preferred

MAC glucuronide linker-1 is a cleavable glucuronide-based linker developed for constructing antibody-drug conjugates (ADCs) in research applications. It is supplied as a solid with reported high purity and specific storage recommendations for stability.

  • Cleavable glucuronide-based linker suitable for ADC construction.
  • Reported purity 96.36% (typical).
  • Molecular weight 897.90 g/mol.
  • Chemical formula C42H47N3O17S.
  • Supplied as a powder; multiple package sizes available.
  • Recommended storage: 4°C under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-02616 Supplier Medchemexpress LLC Supplier No. HY44221500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAC glucuronide linker-1 is a cleavable glucuronide-based linker developed for constructing antibody-drug conjugates (ADCs) in research applications. It is supplied as a solid with reported high purity and specific storage recommendations for stability.

  • Cleavable glucuronide-based linker suitable for ADC construction.
  • Reported purity 96.36% (typical).
  • Molecular weight 897.90 g/mol.
  • Chemical formula C42H47N3O17S.
  • Supplied as a powder; multiple package sizes available.
  • Recommended storage: 4°C under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.