Learn More
Medchemexpress LLC Mac glucuronide linker-1 | 2222981-71-5 | MFCD32671554 | 96.4% | 897.90 | C42H47N3O17S | 50 MG
SDP
Catalog No. 5000202803
Encompass_Preferred
MAC glucuronide linker-1 is a cleavable glucuronide-based linker designed for antibody-drug conjugate (ADC) research. It enables enzymatic release of payloads from antibody-payload conjugates, facilitating controlled drug release in ADC development.
  • Cleavable glucuronide linker for ADCs.
  • Designed for enzymatic payload release by glucuronidase.
  • Suitable for antibody-agent conjugations and linker-payload development.
  • Reported purity approximately 96.4% and molecular weight 897.90.
  • Available in multiple pack sizes, including 50 MG.
Catalog No. 50-002-02803 Supplier Medchemexpress LLC Supplier No. HY4422150MG
MAC glucuronide linker-1 is a cleavable glucuronide-based linker designed for antibody-drug conjugate (ADC) research. It enables enzymatic release of payloads from antibody-payload conjugates, facilitating controlled drug release in ADC development.
  • Cleavable glucuronide linker for ADCs.
  • Designed for enzymatic payload release by glucuronidase.
  • Suitable for antibody-agent conjugations and linker-payload development.
  • Reported purity approximately 96.4% and molecular weight 897.90.
  • Available in multiple pack sizes, including 50 MG.