Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mac glucuronide linker-1 | 2222981-71-5 | MFCD32689543 | 96.4% | 897.90 | C42H47N3O17S | 25 MG
SDP

Catalog No. 5000202045
Encompass_Preferred

MAC glucuronide linker-1 is a cleavable antibody-drug conjugate (ADC) linker intended for research use, designed to release payloads through glucuronide cleavage under target conditions.

  • Cleavable glucuronide-based linker for antibody-drug conjugates.
  • Intended for research use only.
  • CAS number 2222981-71-5.
  • Purity 96.36%.
  • Molecular weight 897.90.
  • Chemical formula C42H47N3O17S.
  • Appearance white to off-white solid.
  • Storage: store under nitrogen; in solvent, -80°C for 6 months and -20°C for 1 month.

Catalog No. 50-002-02045 Supplier Medchemexpress LLC Supplier No. HY4422125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAC glucuronide linker-1 is a cleavable antibody-drug conjugate (ADC) linker intended for research use, designed to release payloads through glucuronide cleavage under target conditions.

  • Cleavable glucuronide-based linker for antibody-drug conjugates.
  • Intended for research use only.
  • CAS number 2222981-71-5.
  • Purity 96.36%.
  • Molecular weight 897.90.
  • Chemical formula C42H47N3O17S.
  • Appearance white to off-white solid.
  • Storage: store under nitrogen; in solvent, -80°C for 6 months and -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.