Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Macamide B | 74058-71-2 | 1 MG
SDP

Catalog No. 5000420337
Encompass_Preferred

Macamide B (N-Benzylhexadecanamide; Macamide 1) is a macamide isolated from Lepidium meyenii. It acts as an inhibitor of fatty acid amide hydrolase (FAAH) and has been shown to exhibit inhibitory effects on FAAH at various concentrations. This product is intended for research purposes only and boasts a high purity.

  • Inhibitor of fatty acid amide hydrolase (FAAH).
  • Exhibits 24.8% and 43.8% inhibition on FAAH at 10 μM and 500 μM, respectively.
  • Used for research purposes only.
  • Has a purity of 99.35%.
  • Storage conditions for powder are at -20°C for 3 years and 4°C for 2 years.
  • Storage conditions in solvent are at -80°C for 6 months and -20°C for 1 month.

Catalog No. 50-004-20337 Supplier Medchemexpress LLC Supplier No. HYN23651MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Macamide B (N-Benzylhexadecanamide; Macamide 1) is a macamide isolated from Lepidium meyenii. It acts as an inhibitor of fatty acid amide hydrolase (FAAH) and has been shown to exhibit inhibitory effects on FAAH at various concentrations. This product is intended for research purposes only and boasts a high purity.

  • Inhibitor of fatty acid amide hydrolase (FAAH).
  • Exhibits 24.8% and 43.8% inhibition on FAAH at 10 μM and 500 μM, respectively.
  • Used for research purposes only.
  • Has a purity of 99.35%.
  • Storage conditions for powder are at -20°C for 3 years and 4°C for 2 years.
  • Storage conditions in solvent are at -80°C for 6 months and -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.