Learn More
NEOGEN MacConkey Agar without Crystal Violet and Salt, 500 g
SDP
Catalog No. 502015363
Encompass_Preferred
MacConkey Agar without Crystal Violet and Salt is used for the isolation and differentiation of Gram-negative enteric bacilli from specimens containing swarming strains of Proteus spp, and is not intended for use in the diagnosis of disease or other conditions in humans.MacConkey Agar is based on the bile salt-neutral red-lactose agar of MacConkey. The original MacConkey medium was used to differentiate strains of Salmonella typhosa from members of the coliform group. Formula modifications improved growth of Shigella and Salmonella strains. These modifications include the addition of 0.5% sodium chloride, decreased agar content, altered bile salts, and neutral red concentrations. The formula modifications improved differential reactions between enteric pathogens and coliforms.For more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar-without-crystal-violet-and-salt
Catalog No. 50-201-5363 Supplier NEOGEN Supplier No. 700004456
MacConkey Agar without Crystal Violet and Salt is used for the isolation and differentiation of Gram-negative enteric bacilli from specimens containing swarming strains of Proteus spp, and is not intended for use in the diagnosis of disease or other conditions in humans.MacConkey Agar is based on the bile salt-neutral red-lactose agar of MacConkey. The original MacConkey medium was used to differentiate strains of Salmonella typhosa from members of the coliform group. Formula modifications improved growth of Shigella and Salmonella strains. These modifications include the addition of 0.5% sodium chloride, decreased agar content, altered bile salts, and neutral red concentrations. The formula modifications improved differential reactions between enteric pathogens and coliforms.For more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar-without-crystal-violet-and-salt