Learn More
Alkali Scientific MacConkey Agar, 500 G
SDP
Catalog No. 5000660404
Encompass_Preferred
MacConkey Agar is a selective medium used for the isolation and differentiation of Gram-negative bacteria. This 500g package is designed for smaller-scale applications, making it suitable for routine microbiological testing and diagnostic purposes. The medium inhibits the growth of Gram-positive organisms while allowing Gram-negative bacteria, particularly enteric pathogens, to grow. Lactose fermenters produce acid and turn the medium pink, while non-fermenters remain colorless. MacConkey Agar is commonly used to detect E. coli, Salmonella, Shigella, and other enteric pathogens in clinical and environmental samples, particularly in food safety testing.
Catalog No. 50-006-60404 Supplier Alkali Scientific Supplier No. ACC003CP
MacConkey Agar is a selective medium used for the isolation and differentiation of Gram-negative bacteria. This 500g package is designed for smaller-scale applications, making it suitable for routine microbiological testing and diagnostic purposes. The medium inhibits the growth of Gram-positive organisms while allowing Gram-negative bacteria, particularly enteric pathogens, to grow. Lactose fermenters produce acid and turn the medium pink, while non-fermenters remain colorless. MacConkey Agar is commonly used to detect E. coli, Salmonella, Shigella, and other enteric pathogens in clinical and environmental samples, particularly in food safety testing.