Learn More
NEOGEN MacConkey Agar, 5 kg
SDP
Catalog No. 502015202
Encompass_Preferred
MacConkey Agar is used for the isolation and differentiation of Gram-negative enteric bacilli in a laboratory setting. Conforms to Harmonized USP/EP/JP Requirements and FDA/BAM. MacConkey Agar is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized USP/EP/JP for isolation and identification of Escherichia coli from non-sterile products. Conforms to USP/EP/JP performance specification. Gelatin serves as source of carbon and nitrogen. Lactose is a fermentable carbohydrate and sodium chloride maintains the osmotic balance. Bile salts and crystal violet act as selective agents inhibiting many Gram-positive bacteria. Escherichia coli can ferment lactose to produce acid which results in a pH drop. This is indicated by neutral red resulting in pink colonies.For more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar
Catalog No. 50-201-5202 Supplier NEOGEN Supplier No. 700003013
MacConkey Agar is used for the isolation and differentiation of Gram-negative enteric bacilli in a laboratory setting. Conforms to Harmonized USP/EP/JP Requirements and FDA/BAM. MacConkey Agar is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized USP/EP/JP for isolation and identification of Escherichia coli from non-sterile products. Conforms to USP/EP/JP performance specification. Gelatin serves as source of carbon and nitrogen. Lactose is a fermentable carbohydrate and sodium chloride maintains the osmotic balance. Bile salts and crystal violet act as selective agents inhibiting many Gram-positive bacteria. Escherichia coli can ferment lactose to produce acid which results in a pH drop. This is indicated by neutral red resulting in pink colonies.For more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar