Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ MacConkey Agar with Ciprofloxacin
SDP

Catalog No. 23001743 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-743 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-743 Supplier Hardy Diagnostics™ Supplier No. G258
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use as a screening medium for the selective isolation of ciprofloxacin-resistant E. coli and other gram-negative rods from clinical samples.

  • Especially useful for screening rectal swab specimens prior to prostate gland biopsy
    • Our media will allow urologists to screen the men first to see if they will be at risk of a difficult to treat infection or septicemia
  • Recommended to be used with a broth enrichment step using BHI
  • Optically clear polystyrene for distorion free viewing
  • Polystyrene Petri plates feature raised rings for stacking ability and to prevent sliding

Specifications

Appearance Clear, Slightly Opalescent
Color Reddish-purple
Content And Storage 2°C to 8°C
Additive Ciprofloxacin
Detects Ciprofloxacin-resistant E. coli, Gram-negative Rods
Form Agar
Format Plate
Media Type MacConkey Agar
Product Type Agar Plate
Target Organism Class Gram-negative Microorganisms
Test Type Antibiotic Susceptibility Sensitivity
Fill Volume 19 mL
Formulation Peptone - 17.0gm, Lactose - 10.0gm, Sodium Chloride - 5.0gm, Proteose Peptone - 3.0gm, Bile Salts - 1.5gm, Neutral Red - 30.0mg, Ciprofloxacin - 10.0mg, Crystal Violet - 1.0mg, Agar - 13.5gm
For Use With (Application) Selective isolation of ciprofloxacin-resistant E. coli and Gram-negative rods
Incubation Conditions 35°C
Incubation Time 24 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Plates, Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Acinetobacter baumannii, Escherichia coli, Enterococcus faecalis, Geobacillus stearothermophilus
Protein Source Meat, Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.