Learn More
Bioworld MacConkey Agar, 500 g
Catalog No. 50254329
Encompass_Preferred
MacConkey Agar, CS For the isolation and differentiation of Gram-negative enteric bacilli from specimens containing swarming strains of Proteus spp.pH: 7.1Working Concentration: 50 g/LDehydrated Appearance: light pinkish-beige, homogeneous powderPrepared Appearance: dark pink-purple agarSoluble in boiling waterContains: Pancreatic Digest of Gelatin Proteose Peptone Lactose Bile Salts Sodium Chloride Crystal Violet Neutral Red Agar
Catalog No. 50-254-329 Supplier Bioworld Supplier No. 306200481
MacConkey Agar, CS For the isolation and differentiation of Gram-negative enteric bacilli from specimens containing swarming strains of Proteus spp.pH: 7.1Working Concentration: 50 g/LDehydrated Appearance: light pinkish-beige, homogeneous powderPrepared Appearance: dark pink-purple agarSoluble in boiling waterContains: Pancreatic Digest of Gelatin Proteose Peptone Lactose Bile Salts Sodium Chloride Crystal Violet Neutral Red Agar