Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MacConkey Agar Medium
GSA_VA

Catalog No. r01552
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
85mm Monoplate
Media Type:
MacConkey Agar Medium
Manufacturing Origin:
United States
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R01552 100/Pk. 85mm Monoplate MacConkey Agar Medium United States
R01550 10/Pk. 85mm Monoplate MacConkey Agar Medium United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01552 Supplier Thermo Scientific™ Supplier No. R01552
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect and enumerate coliform organisms, E. coli, while isolating and detecting Salmonella and Shigella spp. in both pathological and food specimens.

Detect and enumerate coliform organisms, including E. coli, while isolating and detecting Salmonella and Shigella spp. in both pathological and food specimens using Thermo Scientific™ MacConkey Agar.

Urinary tract infections, bacteremia, meningitis, and diarrheal disease are the most frequent clinical syndromes associated with Escherichia coli. Escherichia coli O157:H7 is associated with hemorrhagic colitis and hemolytic-uremic syndrome (HUS). Escherichia coli is an aerobic, gram-negative bacillus most frequently isolated from the intestines of humans and animals. As a result, it is used as an indicator of fecal contamination in water sources. This pathogen readily grows on a variety of culture media including MacConkey Agar.
Salmonella and Shigella are major causes of bacterial enteric illness. Transmission occurs via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water. A relatively small number of organisms ingested is capable of causing symptomatic infection. While multidrug resistant Shigellae are prevalent, a multidrug resistant strain of Salmonella typhimurium (DT104) emerged in 1985 that is resistant to five antibiotics. Salmonella and Shigella are aerobic, gram-negative, oxidase-negative bacilli. Some Salmonella and all Shigella are nonmotile. These organisms grow on standard media, such as MacConkey Agar, SS Agar, and XLD Agar. Enrichment broths should also be used when culturing for these organisms to allow for maximum recovery.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, SMWW, USP.
Color Clear pink
Content And Storage 2°C to 8°C
Format 85mm Monoplate
Manufacturing Origin United States
Media Type MacConkey Agar Medium
Product Type Agar Plate
Fill Volume 19.5 mL
Packaging Type Flow Wrapped - Single Layer
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.