Learn More
HIMEDIA MacConkey Agar, MH081, Microbiology Culture Media, Dehydrated Culture Media, Selective and Differential Agar, USP/EP/BP/JP Microbial Limits, Powder, 500 g
Catalog No. NC4013769
Encompass_Preferred
MacConkey Agar is a selective and differential medium for isolating and differentiating Escherichia coli and other enteric bacteria, formulated for harmonized microbial limit testing per USP, EP, BP and JP. Bile salts and crystal violet suppress Gram-positive flora, lactose with the neutral red indicator distinguishes lactose fermenters as pink colonies from non-fermenters, final pH 7.1. A workhorse of pharmaceutical quality control and clinical enteric bacteriology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013769 Supplier HIMEDIA Supplier No. MH081500G
MacConkey Agar is a selective and differential medium for isolating and differentiating Escherichia coli and other enteric bacteria, formulated for harmonized microbial limit testing per USP, EP, BP and JP. Bile salts and crystal violet suppress Gram-positive flora, lactose with the neutral red indicator distinguishes lactose fermenters as pink colonies from non-fermenters, final pH 7.1. A workhorse of pharmaceutical quality control and clinical enteric bacteriology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.