Learn More
NEOGEN MacConkey Agar No. 2, 500 g
SDP
Catalog No. 502015699
Encompass_Preferred
MacConkey Agar No.2 is a modification of MacConkey Agar which contains bile salts No. 2 for the recognition of Enterococci, and is not intended for use in the diagnosis of disease or other conditions in humans.This agar especially useful when looking for Enterococci in the presence of coliforms and non-lactose fermenters from water, sewage and food products. Enterococci are frequently sought as an index of fecal pollution and appear on this medium as small, intensely colored red-purple colonies. Non-lactose fermenters appear colorless, whilst bile tolerant Gram-positive organisms, such as staphylococci and non-fecal streptococci, are completely inhibited.Formula:Peptone - 20.0 g/LLactose - 10.0 g/LBile Salts No. 2 - 1.5 g/LSodium Chloride - 5.0 g/LNeutral Red - 0.05 g/LCrystal Violet - 0.001 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar-number-2
Catalog No. 50-201-5699 Supplier NEOGEN Supplier No. 700004614
MacConkey Agar No.2 is a modification of MacConkey Agar which contains bile salts No. 2 for the recognition of Enterococci, and is not intended for use in the diagnosis of disease or other conditions in humans.This agar especially useful when looking for Enterococci in the presence of coliforms and non-lactose fermenters from water, sewage and food products. Enterococci are frequently sought as an index of fecal pollution and appear on this medium as small, intensely colored red-purple colonies. Non-lactose fermenters appear colorless, whilst bile tolerant Gram-positive organisms, such as staphylococci and non-fecal streptococci, are completely inhibited.Formula:Peptone - 20.0 g/LLactose - 10.0 g/LBile Salts No. 2 - 1.5 g/LSodium Chloride - 5.0 g/LNeutral Red - 0.05 g/LCrystal Violet - 0.001 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar-number-2