Learn More
NEOGEN MacConkey Agar No. 3, 5 kg
SDP
Catalog No. 502015655
Encompass_Preferred
MacConkey Agar No. 3 is used for the cultivation of Enterobacteriaceae and is not intended for use in the diagnosis of disease or other conditions in humans.A modification recommended by the W.H.O. and the American Public Health Association for the isolation of Enterobacteriaceae from waters and sewage. The medium has been made more selective than MacConkey’s original formula by the use of crystal violet as well as bile salts. Gram positive organisms will not grow on this medium.Formula:Peptone - 20.0 g/LLactose - 10.0 g/LBile Salt No.3 - 1.5 g/LSodium Chloride - 5.0 g/LNeutral Red - 0.03 g/LCrystal Violet - 0.001 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar-number-3
Catalog No. 50-201-5655 Supplier NEOGEN Supplier No. 700004549
MacConkey Agar No. 3 is used for the cultivation of Enterobacteriaceae and is not intended for use in the diagnosis of disease or other conditions in humans.A modification recommended by the W.H.O. and the American Public Health Association for the isolation of Enterobacteriaceae from waters and sewage. The medium has been made more selective than MacConkey’s original formula by the use of crystal violet as well as bile salts. Gram positive organisms will not grow on this medium.Formula:Peptone - 20.0 g/LLactose - 10.0 g/LBile Salt No.3 - 1.5 g/LSodium Chloride - 5.0 g/LNeutral Red - 0.03 g/LCrystal Violet - 0.001 g/LAgar - 15.0 g/LFor more information visit https://www.neogen.com/solutions/Microbiology/macconkey-agar-number-3