Learn More
MOLTOX Molecular Toxicology Inc MacConkey Agar, 20 mL/plate, 10 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948812
Encompass_Preferred
Used for the detection and isolation of gram-negative organisms and for isolating and differentiating lactose fermenting from lactose-nonfermenting enteric bacilli. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-812 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S012.140
Used for the detection and isolation of gram-negative organisms and for isolating and differentiating lactose fermenting from lactose-nonfermenting enteric bacilli. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.