Learn More
Teknova MacConkey Agar Plates. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388029
Encompass
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.
Catalog No. 50-238-8029 Supplier Teknova Supplier No. M0149
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.