Learn More
Teknova MacConkey Agar Plates with 1% Galactose. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387751
Encompass
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.
Catalog No. 50-238-7751 Supplier Teknova Supplier No. M01524PK
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.