Learn More
Teknova MacConkey Agar Plates with Levofloxacin-16. 100mm Plates, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387749
Encompass
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.
Catalog No. 50-238-7749 Supplier Teknova Supplier No. M11524PK
MacConkey agar media aid in the isolation and differentiation of gram-negative bacterial isolates. It serves as an indicator, selective, and differential media, which determines the growth or presence of the gram-negative enteric bacilli from the different samples that are important for the sanitary purpose.