Learn More
MOLTOX Molecular Toxicology Inc MacConkey Agar w/ 10ug/ml Nalidixic Acid, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948827
Encompass_Preferred
Used for the detection and isolation of gram-negative organisms and for isolating and differentiating lactose fermenting from lactose-nonfermenting enteric bacilli. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-827 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S038
Used for the detection and isolation of gram-negative organisms and for isolating and differentiating lactose fermenting from lactose-nonfermenting enteric bacilli. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.