Learn More
Bioworld MacConkey Broth, 500 g
Catalog No. 50254330
Encompass_Preferred
MacConkey BrothFor the detection of coliform bacteria in milk and waterpH: 7.3Working Concentration: 35 g/LPrepared Appearance: dark purple brothSoluble in waterContains: Enzymatic Digest of GelatinLactoseOxbileBromocresol Purple
Catalog No. 50-254-330 Supplier Bioworld Supplier No. 306200491
MacConkey BrothFor the detection of coliform bacteria in milk and waterpH: 7.3Working Concentration: 35 g/LPrepared Appearance: dark purple brothSoluble in waterContains: Enzymatic Digest of GelatinLactoseOxbileBromocresol Purple