Learn More
MOLTOX Molecular Toxicology Inc MacConkey Broth, 100 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948762
Encompass_Preferred
MacConkey broth is utilized as a preliminary test for coliform organisms. It is used for cultivating gram-negative, lactose fermenting bacilli in water and foods. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-762 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26556
MacConkey broth is utilized as a preliminary test for coliform organisms. It is used for cultivating gram-negative, lactose fermenting bacilli in water and foods. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.