Learn More
Alkali Scientific MacConkey / PEA Agar with 5% Sheep Blood, Biplate, 10/Pk
SDP
Catalog No. 5000662842
Encompass_Preferred
MacConkey / PEA Agar with 5% Sheep Blood, Biplate, 10/Pk. This specialized biplate contains MacConkey agar and phenylethyl alcohol (PEA) agar with 5% sheep blood, making it ideal for isolating and differentiating Gram-negative bacteria, including enteric pathogens. The inclusion of blood supports the growth of fastidious organisms, while PEA inhibits Gram-positive bacteria, ensuring clear, distinct colonies for accurate identification.
Catalog No. 50-006-62842 Supplier Alkali Scientific Supplier No. MACBP01
MacConkey / PEA Agar with 5% Sheep Blood, Biplate, 10/Pk. This specialized biplate contains MacConkey agar and phenylethyl alcohol (PEA) agar with 5% sheep blood, making it ideal for isolating and differentiating Gram-negative bacteria, including enteric pathogens. The inclusion of blood supports the growth of fastidious organisms, while PEA inhibits Gram-positive bacteria, ensuring clear, distinct colonies for accurate identification.