Learn More
Bioworld Macerozyme R-10, 5 g
Catalog No. 50198984
Encompass_Preferred
Macerozyme R-10Macerating enzyme from Rhizopus sp. suitable for the isolation of plant cells and can be used in combination with cellulase.Specification:Used in plant protoplast preparation to digest cell wall prior to oranelle isolation.A multi-component enzyme mixture containing approximately: * Cellulase: 0.1 U/mg * Hemicellulase: 0.25 U/mg * Pectinase: 0.5 U/mgSource: Rhizopus sp.Solubility: Soluble in water
Catalog No. 50-198-984 Supplier Bioworld Supplier No. 215600172
Macerozyme R-10Macerating enzyme from Rhizopus sp. suitable for the isolation of plant cells and can be used in combination with cellulase.Specification:Used in plant protoplast preparation to digest cell wall prior to oranelle isolation.A multi-component enzyme mixture containing approximately: * Cellulase: 0.1 U/mg * Hemicellulase: 0.25 U/mg * Pectinase: 0.5 U/mgSource: Rhizopus sp.Solubility: Soluble in water