Learn More
Research Products International Corp Macerozyme R-10, 10 Grams
SDP
Catalog No. 50488794
Encompass_Preferred
Macerating enzyme from Rhizopus sp. Often used in combination with Cellulase (C32200). Optimum pH range 3.5 - 7.0.
Catalog No. 50-488-794 Supplier Research Products International Corp Supplier No. M2201010.0
Macerating enzyme from Rhizopus sp. Often used in combination with Cellulase (C32200). Optimum pH range 3.5 - 7.0.