Learn More
AG Scientific Inc Macerozyme R-10, 5 G
Supplier:  AG Scientific Inc M26755GM
Encompass_Preferred
Also known as Pectinase, Macerozyme R-10 is a macerating enzyme from Rhizopus sp. Often used in combination with Cellulase.CAS Number: 9032-75-1Solubility: 1 mg/mL 0.1 M sodium acetate bufferStorage Temperature: +2-8CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6959