Learn More
Crescent Chemical Co Inc MACEROZYME R-10 FROM RHIZOPUS
SDP
Catalog No. 50247821
Encompass_Preferred
Macerozyme R-10 from Rhizopus sp. lyophil. Macerozyme R-10 is suitable for the isolation of plant cells, and can be used in combination with cellulase »Onozuka R-10« (cat. no. 16419) (1, 2) and with Cellulase »Onozuka RS« (cat. no. 16420). Size - 10G Storage Conditions -
Catalog No. 50-247-821 Supplier Crescent Chemical Co Inc Supplier No. SE28302.03
Macerozyme R-10 from Rhizopus sp. lyophil. Macerozyme R-10 is suitable for the isolation of plant cells, and can be used in combination with cellulase »Onozuka R-10« (cat. no. 16419) (1, 2) and with Cellulase »Onozuka RS« (cat. no. 16420). Size - 10G Storage Conditions -