Learn More
Crescent Chemical Co Inc Macerozyme R-10 from Rhizopus SP, 9032-75-1, MFCD00131809, 2.5G
SDP
Catalog No. 50247820
Encompass_Preferred
Macerozyme R-10 from Rhizopus SP, 9032-75-1, MFCD00131809, 2.5G, Macerozyme R-10 is suitable for the isolation of plant cells, and can be used in combination with cellulase »Onozuka R-10« (cat. no. 16419) (1, 2) and with Cellulase »Onozuka RS«. Storage Conditions: +2 °C TO +8 °C
Catalog No. 50-247-820 Supplier Crescent Chemical Co Inc Supplier No. SE28302.02
Macerozyme R-10 from Rhizopus SP, 9032-75-1, MFCD00131809, 2.5G, Macerozyme R-10 is suitable for the isolation of plant cells, and can be used in combination with cellulase »Onozuka R-10« (cat. no. 16419) (1, 2) and with Cellulase »Onozuka RS«. Storage Conditions: +2 °C TO +8 °C