Learn More
DESERT BIOLOGICALS RESEARCH Macerozyme R10
Catalog No. NC1595511
Encompass_Preferred
Standardized at 3000 maceration units/gram.The activity is based on measuring the volume of single cells released from potato tuber disks. pH: 5.0-6.0
Catalog No. NC1595511 Supplier DESERT BIOLOGICALS RESEARCH Supplier No. 6152
Standardized at 3000 maceration units/gram.The activity is based on measuring the volume of single cells released from potato tuber disks. pH: 5.0-6.0