Learn More
Biocare Medical, LLC Rodent Block R
SDP
Catalog No. 5082928
Encompass_Preferred
Rodent Block R is an immunohistochemical reagent designed for blocking endogenous rat IgG and non-specific background in rat tissues and red blood cells. After the application of Rodent Block R, a mouse monoclonal antibody can be applied. It can also be used on an automated staining system.
Catalog No. 50-829-28 Supplier Biocare Medical, LLC Supplier No. MHRP520H
Rodent Block R is an immunohistochemical reagent designed for blocking endogenous rat IgG and non-specific background in rat tissues and red blood cells. After the application of Rodent Block R, a mouse monoclonal antibody can be applied. It can also be used on an automated staining system.