Learn More
Biocare Medical, LLC MACH 2 Rabbit HRP-Polymer - 6.0 ml
SDP
Catalog No. 5083274
Encompass_Preferred
The conjugated goat anti-rabbit polymer-horseradish peroxidase secondary antibody,reacts with heavy and light chains on rabbit IgG.The new and innovative HRP-polymerization technology provides a significant increase in staining sensitivity when compared to other conventional HRP-conjugated secondary antibodies. Avidin-biotin blocking procedures are not necessary when using the MACH 2 conjugated secondary antibodies.The overall staining procedure uses one less reagent (streptavidin peroxidase) and rinsing step, eliminates avidin-biotin blocking procedures, and may reduce overall staining time. It can be used manually and on automated stainers.
Catalog No. 50-832-74 Supplier Biocare Medical, LLC Supplier No. RHRP520G
The conjugated goat anti-rabbit polymer-horseradish peroxidase secondary antibody,reacts with heavy and light chains on rabbit IgG.The new and innovative HRP-polymerization technology provides a significant increase in staining sensitivity when compared to other conventional HRP-conjugated secondary antibodies. Avidin-biotin blocking procedures are not necessary when using the MACH 2 conjugated secondary antibodies.The overall staining procedure uses one less reagent (streptavidin peroxidase) and rinsing step, eliminates avidin-biotin blocking procedures, and may reduce overall staining time. It can be used manually and on automated stainers.