Learn More
Biocare Medical, LLC MACH 4™ Universal AP Polymer Kit - 6.0 ml
SDP
Supplier:  Biocare Medical, LLC M4U536G
Encompass_Preferred
The MACH 4 Universal AP Polymer Kit detects both mouse and rabbit primary antibodies. This new biotin-free technology uses a specific probe to detect mouse primary antibodies and is then followed by an alkaline phosphatase polymer (AP) that binds to the probe. This new and innovative AP-polymerization technology provides increased staining sensitivity up to 5 to 20 times when compared to Envision and other AP-polymer detection kits. It can be used manually and on automated stainers.Covered by one or more of the following US Pat. Nos. 6,686,461; 6,800,728; 7,102,024; 7,173,125; 7,462,689.
Catalog No. 50-829-13