Learn More
Macherey-Nagel NucleoBond Xtra Combi Rack
Catalog No. NC1664905
Encompass_Preferred
NucleoBond Xtra Combi Rack rack for use with NucleoBond Xtra Midi, Xtra Maxi, Xtra BAC, AX 100, AX 500, AX 2000, and AX 10000 Columns pack of 1
Catalog No. NC1664905 Supplier Macherey-Nagel Supplier No. 740415
NucleoBond Xtra Combi Rack rack for use with NucleoBond Xtra Midi, Xtra Maxi, Xtra BAC, AX 100, AX 500, AX 2000, and AX 10000 Columns pack of 1