Learn More
Shoe Inn LLC Shoe Inn 28g Fabric with Traction Shoe Covers for Shoe Inn Fusion and Stay Dispensers, 1,500 Shoe Covers/750 Pairs, CS
Catalog No. NC1559222
Encompass_Preferred
Our no-slip fabric shoe covers are made of 100% spunbond polypropylene, which provides breathability for non-hazardous and low particulate barrier applications. This low-lint, non-woven fabric helps filter particulates for contamination control in critical environments. These shoe covers have a thin industry standard traction tread pattern on the bottom. Each cartridge contains 100 one-size-fits-most (up to about a size 15 or 16) anti-slip fabric shoe covers. Fifteen (15) cartridges are included in a case for a total of 1,500 shoe covers (750 pairs).
This item is not returnable. View return policy
Catalog No. NC1559222 Supplier Shoe Inn LLC Supplier No. 7FWT100HC
Our no-slip fabric shoe covers are made of 100% spunbond polypropylene, which provides breathability for non-hazardous and low particulate barrier applications. This low-lint, non-woven fabric helps filter particulates for contamination control in critical environments. These shoe covers have a thin industry standard traction tread pattern on the bottom. Each cartridge contains 100 one-size-fits-most (up to about a size 15 or 16) anti-slip fabric shoe covers. Fifteen (15) cartridges are included in a case for a total of 1,500 shoe covers (750 pairs).