Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Machine/Equipment Label, Header: DANGER, Legend: STARTS AUTOMATICALLY

Catalog No. 19110867 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
Header: DANGER, Legend: STARTS AUTOMATICALLY
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19-110-867 Header: DANGER, Legend: STARTS AUTOMATICALLY
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-110-867 Supplier Brady™ Supplier No. 86148
  • $31.25
    $21.65 / Pack of 5

    Save $9.60

    31% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Communicates important safety information and machine operational instructions. Brady Machine and Equipment Labels are made of durable, self-sticking polyester and most feature pictograms as well as text to inform safety procedures and potential hazards with machine operation.

  • Placed on machine or equipment to increase safety
  • Made of B-302 self sticking polyester
  • Available in size (W x H): 12.7 x 8.89cm (5 x 3.5 in.)

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Red, White
Finish Gloss
Material Self-adhesive Polyester (B-302)
Product Type Machine/Equipment Label
Display Text Header: DANGER, Legend: STARTS AUTOMATICALLY
Languages English
Quantity 5/Pk.
Shape Rectangle
Size (English) 5 x 3.5 in. (W x H)
Size (Metric) 127 x 88.9 mm (W x H)
Thickness (English) 0.01 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.