Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Machine/Equipment Label, Header: DANGER, Legend: Wear face shield when using this equipment

Catalog No. 19111849 Shop All Brady Corporation Products
Change view
Click to view available options
Adhesive Type:
Acrylic, Permanent, Cold-temperature
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Adhesive Type
19-111-849 Acrylic, Permanent, Cold-temperature
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-111-849 Supplier Brady™ Supplier No. 86818
Only null left
Add to Cart
Add to Cart

Communicates important safety information and machine operational instructions. Brady Machine and Equipment Labels are made of durable, self-sticking polyester and most feature pictograms as well as text to inform safety procedures and potential hazards with machine operation.

  • Placed on machine or equipment to increase safety
  • Made of B-302 self sticking polyester
  • Available in size (W x H): 12.7 x 8.89cm (5 x 3.5 in.)

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Red, White
Finish Gloss
Material Self-adhesive Polyester (B-302)
Product Type Machine/Equipment Label
Display Text Header: DANGER, Legend: Wear face shield when using this equipment
Languages English
Quantity 5/Pk.
Shape Rectangle
Size (English) 5 x 3.5 in. (W x H)
Size (Metric) 127 x 88.9 mm (W x H)
Thickness (English) 0.01 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.