Learn More
Uline Machine Length Stretch Film Cast 80 gauge
Catalog No. NC2327408
Encompass
Stretch film in continuous roll used for wrapping and unitizing pallet loads, supplied on 3 inch cores, 80 gauge, 20 inches wide, 6000 feet long, providing load containment and protection during shipping and storage.
Catalog No. NC2327408 Supplier Uline Supplier No. S1524
Stretch film in continuous roll used for wrapping and unitizing pallet loads, supplied on 3 inch cores, 80 gauge, 20 inches wide, 6000 feet long, providing load containment and protection during shipping and storage.