Learn More
Uline ****Per Supplier requirements this item has a Minimum order quantity of 6****Machine Length Stretch Film - Cast, 80 gauge, 20" x 5,000'
Catalog No. NC3522215
Encompass
Clear, quiet cast is our best-selling film for uniform loads.80 gauge - For loads up to 2,500 lbs.Smooth, easy release. Wraps loads fast with little effort.Two-sided cling.Stretches up to 250%.
Catalog No. NC3522215 Supplier Uline Supplier No. S2193
Clear, quiet cast is our best-selling film for uniform loads.80 gauge - For loads up to 2,500 lbs.Smooth, easy release. Wraps loads fast with little effort.Two-sided cling.Stretches up to 250%.