Learn More
Uline Adjustable Height Machine Table - 24 x 18 x 18-25""
Catalog No. NC1946549
Encompass
Adjustable Height Machine Table - 24 x 18 x 18-25", Adjustable height tables for light duty drilling and parts assembly.Legs are adjustable in 1" increments.Adjustable bottom shelf.14-gauge steel top and 2" angle legs.
Catalog No. NC1946549 Supplier Uline Supplier No. H6979
Adjustable Height Machine Table - 24 x 18 x 18-25", Adjustable height tables for light duty drilling and parts assembly.Legs are adjustable in 1" increments.Adjustable bottom shelf.14-gauge steel top and 2" angle legs.