Learn More
Grainger Machine Vise 2-1/2 Fixed
Catalog No. 502766227
Encompass_Preferred
The Dayton Machine Vise has a 2-1/2 inch jaw width and opens to a maximum width of 4 inches. It is made from high strength cast iron with a hardened steel vise screw and replaceable serrated steel jaws for gripping. This vise can handle up to 550 lbs of clamping force and mounts to a work surface via the base using bolts. The overall dimensions are 6-1/2 inches tall by 4-3/4 inches wide by 9 inches deep. This is a heavy duty vise well suited for machine shop, garage, or welding applications.
Catalog No. 50-276-6227 Supplier Grainger Supplier No. 16Y478
The Dayton Machine Vise has a 2-1/2 inch jaw width and opens to a maximum width of 4 inches. It is made from high strength cast iron with a hardened steel vise screw and replaceable serrated steel jaws for gripping. This vise can handle up to 550 lbs of clamping force and mounts to a work surface via the base using bolts. The overall dimensions are 6-1/2 inches tall by 4-3/4 inches wide by 9 inches deep. This is a heavy duty vise well suited for machine shop, garage, or welding applications.