Learn More
Grainger Machine Vise 2-7/8 Fixed
Catalog No. 502767006
Encompass_Preferred
The Dayton Machine Vise has a 2-7/8 inch jaw width and opens to a maximum width of 5 inches. It is made from high strength cast iron with a hardened steel vise screw and replaceable steel jaw plates. The vise has a 450 pound clamping force capacity. It can be mounted to a workbench or machine using the included mounting ears and hardware. The overall dimensions are 9 inches high by 7 inches wide by 5-1/2 inches deep. This is a heavy duty vise suitable for clamping metal workpieces during machining operations.
Catalog No. 50-276-7006 Supplier Grainger Supplier No. 16Y480
The Dayton Machine Vise has a 2-7/8 inch jaw width and opens to a maximum width of 5 inches. It is made from high strength cast iron with a hardened steel vise screw and replaceable steel jaw plates. The vise has a 450 pound clamping force capacity. It can be mounted to a workbench or machine using the included mounting ears and hardware. The overall dimensions are 9 inches high by 7 inches wide by 5-1/2 inches deep. This is a heavy duty vise suitable for clamping metal workpieces during machining operations.