Learn More
Grainger Machine Vise 2 Fixed
Catalog No. 502767164
Encompass_Preferred
The Dayton Machine Vise has a 2 inch jaw face and a maximum jaw opening of 2 inches. It is made of ductile iron with a hardened steel vise screw and replaceable steel jaw plates. The vise has a flat base that allows it to be permanently mounted. The vise can handle up to 550 lbs of clamping force. This medium duty machine vise is suitable for precision workholding applications.
Catalog No. 50-276-7164 Supplier Grainger Supplier No. 16Y479
The Dayton Machine Vise has a 2 inch jaw face and a maximum jaw opening of 2 inches. It is made of ductile iron with a hardened steel vise screw and replaceable steel jaw plates. The vise has a flat base that allows it to be permanently mounted. The vise can handle up to 550 lbs of clamping force. This medium duty machine vise is suitable for precision workholding applications.