Learn More
Grainger Ken Forging Machinery Eye Bolt, Shoulder, 304 Stainless Steel, 3/4-10 Thread, 2 in. Shank, 1 ea
Catalog No. 502780865
Encompass_Preferred
This drop forged machinery eye bolt with shoulder is designed for heavy-duty lifting applications. Manufactured from 304 stainless steel with an 18-8 grade, it is passivated and electropolished for enhanced corrosion resistance and durability. It features a 3/4 in.-10 UNC-2A thread, a 2 in. shank length, and a 5,000 lb. working load limit.
  • Shoulder pattern design
  • Constructed from 304 stainless steel (ASTM A193-B8)
  • Passivated and electropolished finish
  • 3/4 in.-10 UNC-2A thread size
  • 5,000 lb. working load limit
  • 1-1/2 in. eye inside diameter
  • 2-13/16 in. eye outside diameter
  • ASME B18.15 dimensional standards
Catalog No. 50-278-0865 Supplier Grainger Supplier No. 19L126
This drop forged machinery eye bolt with shoulder is designed for heavy-duty lifting applications. Manufactured from 304 stainless steel with an 18-8 grade, it is passivated and electropolished for enhanced corrosion resistance and durability. It features a 3/4 in.-10 UNC-2A thread, a 2 in. shank length, and a 5,000 lb. working load limit.
  • Shoulder pattern design
  • Constructed from 304 stainless steel (ASTM A193-B8)
  • Passivated and electropolished finish
  • 3/4 in.-10 UNC-2A thread size
  • 5,000 lb. working load limit
  • 1-1/2 in. eye inside diameter
  • 2-13/16 in. eye outside diameter
  • ASME B18.15 dimensional standards