Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM2348P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: D11. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Liver or Histiocytoma. Cellular Staining: Cell Surface & Cytoplasmic. Buffer: 10mM PBS with 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Membrane preparation from human hepatocytes. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5451