Learn More
Medchemexpress LLC HY-12426A 5mg Medchemexpress, mAChR-IN-1 (hydrochloride) CAS:119391-73-0 Purity:>98%
SDP
Catalog No. 501932591
Encompass_Preferred
Medchemexpress, HY-12426A 5mg mAChR-IN-1 (hydrochloride) CAS:119391-73-0 mAChR-IN-1 hydrochloride is a potent muscarinic cholinergic receptor (mAChR) antagonist, with an IC 50 of 17 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2591 Supplier Medchemexpress LLC Supplier No. HY12426A5MG
Medchemexpress, HY-12426A 5mg mAChR-IN-1 (hydrochloride) CAS:119391-73-0 mAChR-IN-1 hydrochloride is a potent muscarinic cholinergic receptor (mAChR) antagonist, with an IC 50 of 17 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.