Learn More
Medchemexpress LLC mAChR-IN-1 hydrochloride | 119391-73-0 | 99.6% | 524.82 g/mol | C23H26ClIN2O2 | 100MG
SDP
Catalog No. 5000238938
Encompass_Preferred
mAChR-IN-1 hydrochloride is a research-grade muscarinic acetylcholine receptor antagonist provided for laboratory use. It exhibits potent inhibitory activity against mAChR with a reported IC50 of 17 nM. The compound is supplied as a hydrochloride salt, with formula C23H26ClIN2O2 and molecular weight 524.82 g/mol.
Catalog No. 50-002-38938 Supplier Medchemexpress LLC Supplier No. HY12426A100MG
mAChR-IN-1 hydrochloride is a research-grade muscarinic acetylcholine receptor antagonist provided for laboratory use. It exhibits potent inhibitory activity against mAChR with a reported IC50 of 17 nM. The compound is supplied as a hydrochloride salt, with formula C23H26ClIN2O2 and molecular weight 524.82 g/mol.