Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC mAChR-IN-1 (hydrochloride) | 119391-73-0 | 99.6% | 524.82 g/mol | C23H26ClIN2O2 | 10MM 1ML
SDP

Catalog No. 5000238645
Encompass_Preferred

mAChR-IN-1 hydrochloride is a potent muscarinic cholinergic receptor antagonist (IC50 17 nM) provided for research use. It is offered as a solid and as a pre-made solution in DMSO, intended for in vitro pharmacology and receptor binding studies.

  • Potent mAChR antagonist (IC50 17 nM).
  • High purity: 99.6% (LCMS).
  • Chemical formula: C23H26ClIN2O2.
  • Molecular weight: 524.82 g/mol.
  • Physical appearance: white to off-white solid.
  • Available as solids (5-100 mg) and 10 mM solution in DMSO, 1 mL.
  • Supplied for research use only.

Catalog No. 50-002-38645 Supplier Medchemexpress LLC Supplier No. HY12426A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

mAChR-IN-1 hydrochloride is a potent muscarinic cholinergic receptor antagonist (IC50 17 nM) provided for research use. It is offered as a solid and as a pre-made solution in DMSO, intended for in vitro pharmacology and receptor binding studies.

  • Potent mAChR antagonist (IC50 17 nM).
  • High purity: 99.6% (LCMS).
  • Chemical formula: C23H26ClIN2O2.
  • Molecular weight: 524.82 g/mol.
  • Physical appearance: white to off-white solid.
  • Available as solids (5-100 mg) and 10 mM solution in DMSO, 1 mL.
  • Supplied for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.