Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC mAChR-IN-1 hydrochloride | 119391-73-0 | 99.6% | 524.82 g/mol | C23H26ClIN2O2 | 25MG
SDP

Catalog No. 5000238643
Encompass_Preferred

mAChR-IN-1 hydrochloride is a research-grade small-molecule antagonist of muscarinic cholinergic receptors (mAChR). It exhibits potent inhibition of mAChR (reported IC50 17 nM) and is supplied as the hydrochloride salt for use in biochemical and pharmacological studies.

  • Potent mAChR antagonist with an IC50 of 17 nM.
  • High chemical purity (99.6%).
  • Molecular weight 524.82 g/mol and formula C23H26ClIN2O2.
  • Available in multiple pack sizes, including a 25 MG option.
  • Supplied as the hydrochloride salt for improved solubility.
  • Suitable for in vitro pharmacology and receptor profiling.
  • Provided with supporting product documentation and datasheet.

Catalog No. 50-002-38643 Supplier Medchemexpress LLC Supplier No. HY12426A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

mAChR-IN-1 hydrochloride is a research-grade small-molecule antagonist of muscarinic cholinergic receptors (mAChR). It exhibits potent inhibition of mAChR (reported IC50 17 nM) and is supplied as the hydrochloride salt for use in biochemical and pharmacological studies.

  • Potent mAChR antagonist with an IC50 of 17 nM.
  • High chemical purity (99.6%).
  • Molecular weight 524.82 g/mol and formula C23H26ClIN2O2.
  • Available in multiple pack sizes, including a 25 MG option.
  • Supplied as the hydrochloride salt for improved solubility.
  • Suitable for in vitro pharmacology and receptor profiling.
  • Provided with supporting product documentation and datasheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.